Merck
All Photos(2)

GF99984335

Gadolinium

rod, 25mm, diameter 12.5mm, cast, 99%

Synonym(s):
Gadolinium, GD007925
Linear Formula:
Gd
CAS Number:
Molecular Weight:
157.25
MDL number:
PubChem Substance ID:
NACRES:
NA.23

assay

≥99%

form

rod

manufacturer/tradename

Goodfellow 999-843-35

resistivity

126 μΩ-cm, 20°C

bp

3273 °C (lit.)

mp

1313 °C (lit.)

density

7.886 g/mL at 25 °C (lit.)

SMILES string

[Gd]

InChI

1S/Gd

InChI key

UIWYJDYFSGRHKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Water-react 3

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service