Merck
All Photos(1)

THS06146

ThalesNano MicroCatCart catalyst cartridge system

L × diam. 30 mm × 4 mm, 5% Ir/CaCO3

Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
NACRES:
NB.51

manufacturer/tradename

Nanotechnologiai Rt.

L × diam.

30 mm × 4 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

MicroCatCart is a trademark of ThalesNano Nanotechnology Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service