All Photos(1)

Z105740

PTFE stirrer blade

fits shaft o.d., 10 mm (with button)

NACRES:
NB.72

material

PTFE

feature

fits shaft o.d. 10 mm (with button)
for flask size 5,000 mL

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F37973-0000

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service