Merck
All Photos(1)

C-237

Supelco

Chloroquine diphosphate solution

1 mg/mL (Methanol:Water (90:10) (v/v), (as free base)), certified reference material, ampule of 1 mL, Cerilliant®

Empirical Formula (Hill Notation):
C18H26ClN3 · 2H3PO4
CAS Number:
Molecular Weight:
515.86

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (Methanol:Water (90:10) (v/v), (as free base))

format

single component solution

storage temp.

−20°C

SMILES string

O=P(OO)=O.ClC1=CC=C2C(N=CC=C2NC(CCCN(CC)CC)C)=C1.O=P(OO)=O.[HH].[HH]

General description

Chloroquine, sold under trade names such as Aralen, Avloclor among others, is an aminoquinoline that has antimalarial and anticancer activities. Chloroquine is being studied to treat COVID-19 as of 2020. A Snap-N-Spike® reference solution applicable for use in pharma research and clinical toxicology analysis by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Cerilliant Corporation

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service