1.00156
BP
JP
NF
Ph. Eur.
2.6 mmHg ( 20 °C)
EMPROVE® ESSENTIAL
99.5-100.5% dry basis (alkalimetric)
crystalline
solid
3.8-4.8 (20 °C, 33 g/L in H2O)
160 °C (dec.) (lit.)
49.2 g/L
1.489 g/cm3 at 23 °C
sulfate (SO42-): ≤0.04%
As: ≤0.0005%
heavy metals (as Pb): ≤0.0010%
liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
2-30°C
OB(O)O
1S/BH3O3/c2-1(3)4/h2-4H
KGBXLFKZBHKPEV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Danger
Repr. 1B
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK 1
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Quality (COQ).
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials.
Free of charge - Sign in to download
Available by subscription or for a fee
Available by subscription or for a fee
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service