Merck
  • 09684
All Photos(1)

09684

Sigma-Aldrich

Ammonia

puriss., anhydrous, ≥99.9%

Synonym(s):
Ammonia gas
Linear Formula:
NH3
CAS Number:
Molecular Weight:
17.03
Beilstein:
3587154
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

vapor density

0.6 (vs air)

Quality Level

vapor pressure

8.75 atm ( 21 °C)

grade

puriss.

assay

≥99.9%

autoignition temp.

1203 °F

expl. lim.

25 %

impurities

≤0.01% water

bp

−33 °C (lit.)

mp

−78 °C (lit.)

SMILES string

N

InChI

1S/H3N/h1H3

InChI key

QGZKDVFQNNGYKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Cylinder with net ~5.5 kg
DIN 477 nr. 6 (W 21.80 X 1/14" right handed thread)

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Press. Gas Liquefied gas - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service