All Photos(1)

26614

Sigma-Aldrich

Hydrogen chloride

puriss., ≥99.8%

Linear Formula:
HCl
CAS Number:
Molecular Weight:
36.46
Beilstein:
1098214
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

vapor density

1.3 (vs air)

vapor pressure

613 psi ( 21.1 °C)

grade

puriss.

assay

≥99.8%

impurities

≤10 ppm Cl2
≤10 ppm water

bp

−85 °C (lit.)

mp

−114 °C (lit.)

density

1.2 g/mL at 25 °C (lit.)

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Cylinder with net ~10L
DIN 477 nr.8 (W 1" right handed thread)

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Press. Gas Compr. Gas - Skin Corr. 1A

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Sonam Dilwali et al.
Cancer biology & therapy, 16(1), 170-175 (2015-02-19)
Vestibular schwannoma (VS), the fourth most common intracranial tumor, arises from the Schwann cells of the vestibular nerve. Although several pathways have been independently implicated in VS pathobiology, interactions among these pathways have not been explored in depth. We have
Brian W Simons et al.
The Journal of pathology, 235(3), 478-489 (2014-10-29)
Inflammation is associated with several diseases of the prostate including benign enlargement and cancer, but a causal relationship has not been established. Our objective was to characterize the prostate inflammatory microenvironment after infection with a human prostate-derived bacterial strain and
Jingjing Cao et al.
Journal of virology, 89(5), 2777-2791 (2014-12-30)
Microtubule transport of circovirus from the periphery of the cell to the nucleus is essential for viral replication in early infection. How the microtubule is recruited to the viral cargo remains unclear. In this study, we observed that circovirus trafficking
Sonali Nashine et al.
Investigative ophthalmology & visual science, 56(1), 221-231 (2014-11-22)
To investigate the role of C/EBP homologous protein (CHOP), a proapoptotic protein, and the unfolded protein response (UPR) marker that is involved in endoplasmic reticulum (ER) stress-mediated apoptosis in mouse retinal ganglion cell (RGC) death following ischemia/reperfusion (I/R) injury. Retinal
Elizabeth A Ennis et al.
ACS chemical neuroscience, 6(3), 417-427 (2015-01-07)
The high-affinity choline transporter (CHT) is the rate-limiting determinant of acetylcholine (ACh) synthesis, yet the transporter remains a largely undeveloped target for the detection and manipulation of synaptic cholinergic signaling. To expand CHT pharmacology, we pursued a high-throughput screen for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service