Merck
All Photos(1)

33765

Supelco

(Z)-Mevinphos solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Synonym(s):
(Z)-3-(Dimethoxyphosphinyloxy)-2-butenoic acid methyl ester, trans-Mevinphos solution
Empirical Formula (Hill Notation):
C7H13O6P
CAS Number:
Molecular Weight:
224.15
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

COC(=O)\C=C(\C)OP(=O)(OC)OC

InChI

1S/C7H13O6P/c1-6(5-7(8)10-2)13-14(9,11-3)12-4/h5H,1-4H3/b6-5-

InChI key

GEPDYQSQVLXLEU-WAYWQWQTSA-N

Related Categories

General description

(Z)-Mevinphos is the Z-isomer of the organophosphorus insecticide, mevinphos, used to control chewing and sucking insect pests in crop production. Its mode of action involves the inhibition of acetylcholinesterase activity in insect pests.

Application

(Z)-Mevinphos may be used as an analytical reference standard for the determination of the analyte in groundwater samples using automated online liquid-solid extraction followed by liquid chromatography-atmospheric pressure chemical ionization mass spectrometry (LC/APCI-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service