Merck
All Photos(1)

65012BC

Millipore

mEHEC®

BioControl, Dehydrated powder culture medium for selectively enriching Enterohemorrhagic E. coli O157:H7 and Salmonella in selected foods

Synonym(s):
Modified Enterohemorrhagic Escherichia coli Broth

sterility

non-sterile

form

powder

shelf life

6‑36 mo.

manufacturer/tradename

BioControl Systems 65012-10K
BioControl Systems 65012-500

storage condition

dry media

technique(s)

microbiological culture: suitable

application(s)

food and beverages
pathogen testing
sample preparation

compatibility

for use with Assurance GDS® Tq from BioControl Systems, Inc.
for use with TRANSIA® AG (4000BC)
for use with TRANSIA® AG (61001BC)
for use with TRANSIA® AG (67000BC)
for use with VlP® Gold (60036BC)
for use with VlP® Gold (60037BC)
for use with VlP® Gold (60038BC)

storage temp.

room temp

suitability

Salmonella spp.
enterovirulent E. coli (EEC)

General description

mEHEC® Broth is a dehydrated culture medium (in powder form) used for selective enrichment of E. coli O157:H7,E. coli non-O157:H7 STEC, and Salmonella spp. in selected food and environmental samples.

Application

Dehydrated culture medium for selective enrichment of pathogenic bacteria in selected food and environmental samples. For use with Assurance® GDS, TRANSIA Plate, TRANSIA AG, and VIP® Gold.

Components

1 bucket containing 10 Kg of dehydrated mEHEC media (65012-10K) or 1 x 500 g container of dehydrated mEHEC media (65012-500)

Legal Information

Assurance is a registered trademark of BioControl Systems, Inc.
TRANSIA is a trademark of BioControl Systems, Inc.
VIP is a registered trademark of BioControl Systems, Inc.
mEHEC is a registered trademark of BioControl Systems, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service