Skip to Content
Merck

945528

Copper(I) bromide

≥99.5%, purum

Synonym(s):

Cu(I)Br, CuBr, Cuprous bromide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
CuBr
CAS Number:
Molecular Weight:
143.45
UNSPSC Code:
12352324
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

purum

Quality Level

assay

≥99.5%

form

powder or chunks

reaction suitability

reagent type: catalyst
core: copper
reaction type: click chemistry

color

white to green

mp

504 °C (lit.)

density

4.71 g/mL at 25 °C (lit.)

SMILES string

[Cu]Br

InChI

1S/BrH.Cu/h1H;/q;+1/p-1

InChI key

NKNDPYCGAZPOFS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Copper(I) bromide (CuBr) is an inorganic salt commonly used as a catalyst or a source of Cu(I) in organic synthesis.

Application

Copper(I) bromide has been used in:
  • The synthesis of tertiary alkyl amines via photoinduced copper catalysed nucelophilic substitution
  • The synthesis of complex secondary amines via photoredox-catalyzed four component click reactions
  • The synthesis of mechanophore polymers that rupture (C-C) via heterolytic, homolytic and concerted pathways
  • The intramolecular annulation of diaryl cyclic chloranes and bromanes

Features and Benefits

This high quality copper(I) bromide is tested to be extremely low in iron impurities. It is suitable for use in further copper mediated synthesis and catalysis, especially in copper mediated azide-alkyne cycloaddition click chemistry (CuAAC).

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
945528-100G04065273688344
945528-1KG04065273688351

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service