Merck
All Photos(1)

ARK2197

SAFC

Sulphuric Acid

new

0.5 M

Synonym(s):
Sulfuric acid
Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:

vapor density

<0.3 (25 °C, vs air)

Quality Level

vapor pressure

1 mmHg ( 146 °C)

concentration

0.5 M

bp

~290 °C (lit.)

density

1.840 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service