Merck
All Photos(1)

PEA1903

Supelco

Sulfuric Acid in Impinger Solution

Proficiency Testing Material

EC Number:
NACRES:
NA.24

Quality Level

Agency

EPA (Method 8)

packaging

ampule of 2 × 20 mL

concentration

in water

application(s)

environmental

format

matrix material

General description

Sulfuric Acid in Impinger Solution proficiency testing material is produced in accordance with ISO/IEC 17043:2010. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies, or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. Our Air Quality & Emissions Proficiency Testing Samples are designed for use with common methods for monitoring and measuring the air quality and are available in a variety of sampling matrices such as Impinger solutions, Filter paper, PUF cartridges and Absorbent tubes.
These samples are typically provided in duplicate for use in case of lab mishap, for QC purposes or to analyze multiple methods.

Type of PT Scheme: Air Quality and Emissions

Matrix: Impinger Solution

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of Sulfuric Acid in Impinger Solution can be found in the reporting packet which will be available to download at PT Portal.

Application

Sulfuric Acid in Impinger Solution is an air PT standard, suitable for analyzing sulfuric acid in impinger solution.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team.

Features and Benefits

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Preparation Note

Description

  • The PT sample is packaged as a 20 mL concentrate.
  • The sample matrix is water.
  • This PT is suitable for EPA Method 8.

Sample Preparation

  • To prepare the solution, partially fill a 1000 mL volumetric flask with reagent-grade water (near 20 °C).
  • Open ampule PEA1903-20ML (be careful to avoid cuts on glass) and transfer 10.0 mL of the concentrate into the volumetric flask.
  • Fill to volume (1000 mL) with reagent-grade water and mix well.
  • The sample is now ready for immediate analysis.
  • For calculation purposes, assume 1.0 dscm of air.

Storage and Stability

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Other Notes

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

Proficiency Testing Schedule (2021-2022)


Air
AIR21-2 April 7 - May 21, 2021
AIR21-4 October 6 - November 19, 2021

AIR22-2 April 6 - May 20, 2022
AIR22-4 October 5 - November 18, 2022

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Sulfuric acid 10-150 mg/dscm

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service