Merck
All Photos(1)

SA0190BC

Millipore

TRANSIA PLATE Salmonella Gold 10X

BioControl, ELISA test for detection of Salmonella in food and environmental samples

Synonym(s):
TPSG

Agency

AFNOR

shelf life

3‑9 mo.

packaging

pkg of 960 tests

manufacturer/tradename

BioControl Systems SA0190
TRANSIA

technique(s)

ELISA: suitable

input

food(s)

application(s)

environmental
food and beverages
pathogen testing

storage temp.

2-8°C

suitability

Salmonella spp.

General description

TRANSIA PLATE Salmonella Gold is an ELISA test kit. It incorporates proven antibody-antigen "sandwich" assay technology for the detection of Salmonella species in both food and environmental samples. Highly specific antibodies coated to polystyrene microtiter plate wells capture and bind the target antigen, if present. A detection antibody linked to a conjugate enzyme is introduced to form an antibody/antigen/antibody sandwich. Finally, a substrate is added and converted by the conjugate enzyme to produce a color change indicating the presence of the target pathogen. The extensive AOAC and AFNOR validations and built-in positive and negative controls combine to provide users with the necessary confidence in the results. The efficient divisible 96-well format and available automation make it an ideal solution for high volume testing.

Application

TRANSIA PLATE Salmonella Gold is intended for the detection of Salmonella spp. The assay reliably recovers and detects low levels of Salmonella in a variety of food and environmental samples.

Features and Benefits

  • Accurate Method for Salmonella Detection:

TRANSIA PLATE Salmonella Gold is a sandwich enzyme immunoassay for the detection of Salmonella in food, feed and environmental samples. The test utilizes highly specific proprietary antibodies to create an antigen antibody complex which, if present, creates a color change reaction upon addition of the substrate which is read with a microplate reader.

  • Standardized Protocol – Results in 24 hours:

The same enrichment media and incubation time for all samples reduces media preparation time and increases lab efficiency. Enrichment steps are harmonized with ISO reference method and are common for all matrices.

  • Automation Ready:

TRANSIA Plate Salmonella Gold can be fully automated with the Gemini® microplate processor.

  • Extensively validated

TRANSIA Plate Salmonella Gold has numerous validations covering all foods, feed and environmental samples from organizations including AFNOR and AOAC Research Institute.

Components

  • Microtiter plate with divisible strips, 96 wells (8 wells x 12 strips) – 10 pc
  • Lid for the microtiter plate – 1 pc
  • Negative control – ready to use – 2 x 13 mL
  • Positive control – ready to use – 2 x 8 mL
  • Washing buffer – concentrated 20X – 1 x 800 mL
  • Conjugate: anti-Salmonella antibodies conjugated to peroxydase – ready to use – 2 x 57 mL
  • Substrate: TMB – ready to use – 2 x 57 mL
  • Stop solution: H2SO4 – ready to use – 2 x 57 mL
  • Extraction reagent – ready to use – 2 x 60 mL

Testing Method

ELISA

Legal Information

Gemini is a registered trademark of Stratec
TRANSIA is a trademark of BioControl Systems, Inc.

Kit Components Also Available Separately

Product No.
Description
SDS

  • Transia Plate Salmonella Gold Substrate, Filled 57 mL

  • Transia Plate Extraction Reagent, Filled 60 mL, 500 test

  • Wash Buffer, 800 mL fill, 960 test

  • Stop Solution, 57 mL, 960 test

  • Transia Plate Salmonella Gold Coated Plate

  • Transia Plate Salmonella Gold Conjugate, Filled 57 mL

  • Transia Plate Salmonella Gold Positive Control, Filled 8 mL

  • Transia Plate Salmonella Gold Negative Control, Filled 10 mL

See All (8)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service