Merck
All Photos(2)

MAK216

Sigma-Aldrich

High Sensitivity NADPH Assay Kit

Sufficient for 100 Fluorometric tests

NACRES:
NA.84

detection method

fluorometric

storage temp.

−20°C

General description

The reduced form of nicotinamide dinucleotide (NADPH) functions as the cofactor for many redox enzymes and contributes to reductive biosynthetic pathways. NADPH is mainly produced from glucose through the oxidative pentose phosphate pathway.

Application

High Sensitivity NADPH Assay Kit has been used to determine the intracellular NAD(P)H and NAD(P)+ levels.

Suitability

Suitable for the detection of NADPH in a variety of biological samples such as cell and tissue culture supernatants and purified mitochondria.

Principle

The High Sensitivity NADPH Quantification Fluorometric Assay Kit is suitable for quantifying NADPH (ranging from 2-10 pmole/well) in tissues, cells, and mitochondria. The kit detects intracellular NAPDH in the enzyme recycling reaction and does not require purified NADPH from the samples. NADPH is determined by measuring a fluorescent product (λex = 535/ λem = 587 nm) proportional to the amount of NADPH present.

Kit Components Only

Product No.
Description

  • NADPH Extraction Buffer

  • NADPH Cycling Buffer

  • High-Sensitivity Probe

  • NADPH Cycling Enzyme Mix

  • NADPH Standard

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Product Information Sheet

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service