Merck
All Photos(1)

150692

Supelco

Sulfuric acid solution

0.25 M, for 3S adapter technology

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:

assay

0.24875-0.25125 M

form

liquid

reaction suitability

reaction type: Acid-base reactions

concentration

0.25 M

technique(s)

titration: suitable

volume

1 L

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

3S - Safe, Smart, Secure
An intelligent packaging solution protected with originality seal.
Reagent information is transferred via RFID from reagent bottle to titrator.
3S reagents are compatible only with the Metrohm Omnis titration platform.

Please see here SigmaAldrich.com/3S

Other Notes

Once the bottle is opened, storage time is limited.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Related Content

3S Reagents for Volumetric Titration

3S is an intelligent packaging solution protected with an originality seal. The cap provides information via Radio-frequency identification or RFID, enabling automated data transfer between the reagent and titration instrument. Intelligent communication for improved performance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service