Merck
All Photos(1)

150696

Supelco

Hydrochloric acid solution

1 M, for 3S adapter technology

Synonym(s):
Hydrogen chloride solution
Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:

vapor pressure

190 mmHg ( 25 °C)

form

liquid

reaction suitability

reaction type: Acid-base reactions

concentration

0.995-1.005 M
1 M

technique(s)

titration: suitable

volume

1 L

bp

100 °C (lit.)

density

1 g/cm3

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Compare Similar Items

View Full Comparison

1 of 4

This Item
W5305742104150677
Hydrochloric acid solution 1 M, for 3S adapter technology

Supelco

150696

Hydrochloric acid solution

Hydrochloric acid solution volumetric, 0.1 M HCl (0.1N), endotoxin free

Supelco

2104

Hydrochloric acid solution

Hydrochloric acid solution 0.1 M, for 3S adapter technology

Supelco

150677

Hydrochloric acid solution

reaction suitability

reaction type: Acid-base reactions

reaction suitability

-

reaction suitability

-

reaction suitability

reaction type: Acid-base reactions

technique(s)

titration: suitable

technique(s)

-

technique(s)

-

technique(s)

titration: suitable

vapor pressure

190 mmHg ( 25 °C)

vapor pressure

190 mmHg ( 25 °C)

vapor pressure

190 mmHg ( 25 °C)

vapor pressure

190 mmHg ( 25 °C)

form

liquid

form

liquid

form

liquid

form

liquid

concentration

0.995-1.005 M

concentration

32 wt. % in H2O

concentration

0.1 M HCl (0.1N)

concentration

0.0995-0.1005 M, 0.1 M

volume

1 L

volume

-

volume

-

volume

1 L

General description

3S - Safe, Smart, Secure
An intelligent packaging solution protected with originality seal.
Reagent information is transferred via RFID from reagent bottle to titrator.
3S reagents are compatible only with the Metrohm Omnis titration platform.

Please see here SigmaAldrich.com/3S

Other Notes

Once the bottle is opened, storage time is limited.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service