Merck
All Photos(1)

40095-U

Supelco

Chiral Test Mix for Astec® CHIROBIOTIC®

analytical standard

Synonym(s):
5-Methyl-5-phenylhydantoin
Empirical Formula (Hill Notation):
C10H10N2O2
CAS Number:
Molecular Weight:
190.20
MDL number:
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

packaging

ampule of 1 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

199-201 °C (lit.)

format

multi-component solution

separation technique

chiral

storage temp.

2-8°C

SMILES string

CC1(NC(=O)NC1=O)c2ccccc2

InChI

1S/C10H10N2O2/c1-10(7-5-3-2-4-6-7)8(13)11-9(14)12-10/h2-6H,1H3,(H2,11,12,13,14)

InChI key

JNGWGQUYLVSFND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Sigma-Aldrich Co. LLC
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    5-Methyl-5-phenylhydantoin 5000 μg/mL

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service