Merck
All Photos(1)

47414

Supelco

EPA 8240B/8260A GC-MS tuning mix

certified reference material, 25 μg/mL in methanol

Synonym(s):
4-Bromofluorobenzene
Empirical Formula (Hill Notation):
C6H4BrF
Molecular Weight:
175.00
EC Number:

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 8240,8260

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL
pkg of 4 ampules

concentration

25 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

1 of 4

This Item
4707747788Z569879
grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

analytical standard

grade

-

packaging

ampule of 1 mL, pkg of 4 ampules

packaging

ampule of 1 mL

packaging

ampule of 1 mL

packaging

-

concentration

25 μg/mL in methanol

concentration

25 μg/mL in methanol

concentration

in methyl tert-butyl ether (varied)

concentration

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

application(s)

environmental

application(s)

environmental

application(s)

agriculture
environmental

application(s)

-

format

multi-component solution

format

multi-component solution

format

multi-component solution

format

-

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    4-Bromofluorobenzene

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service