Merck
All Photos(1)

506524

Supelco

EPA 524 Rev 4 Update Mix

200 μg/mL each component in methanol, analytical standard

Synonym(s):
EPA 524.2 Rev 4 Update Mix
EC Number:

grade

analytical standard

Quality Level

Agency

EPA 524.1,524.2

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

200 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Acrylonitrile
    Allyl chloride
    tert-Butyl methyl ether
    Carbon disulfide
    Chloroacetonitrile
    1-Chlorobutane
    trans-1,4-Dichloro-2-butene
    Diethyl ether
    Ethyl methacrylate
    Hexachloroethane
    Methacrylonitrile
    Methyl acrylate
    Methyl methacrylate
    Nitrobenzene
    2-Nitropropane
    Pentachloroethane
    Propionitrile
    Tetrahydrofuran
    See All (18)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service