Merck
All Photos(2)

V900279

Sigma-Aldrich

Strontium chloride hexahydrate

Vetec, reagent grade, 99%

Linear Formula:
SrCl2 · 6H2O
CAS Number:
Molecular Weight:
266.62
EC Number:
MDL number:
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

99%

form

chunks or granules
crystalline powder
powder or crystals

mp

115 °C (lit.)

SMILES string

O.O.O.O.O.O.Cl[Sr]Cl

InChI

1S/2ClH.6H2O.Sr/h2*1H;6*1H2;/q;;;;;;;;+2/p-2

InChI key

AMGRXJSJSONEEG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service