Merck
All Photos(3)

V900438

Sigma-Aldrich

L-Lysine monohydrochloride

Vetec, reagent grade, ≥98%

Synonym(s):
(S)-2,6-Diaminohexanoic acid monohydrochloride
Linear Formula:
H2N(CH2)4CH(NH2)CO2H·HCl
CAS Number:
Molecular Weight:
182.65
Beilstein:
3563889
EC Number:
MDL number:
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

≥98%

form

powder

color

white to off-white

mp

263 °C (dec.) (lit.)

SMILES string

OC([C@@H](N)CCCCN)=O.[H]Cl

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1

InChI key

BVHLGVCQOALMSV-JEDNCBNOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

100, 500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service