Merck
All Photos(3)

V900641

Sigma-Aldrich

DL-Alanine

Vetec, reagent grade, 98%

Synonym(s):
(±)-2-Aminopropionic acid
Linear Formula:
CH3CH(NH2)COOH
CAS Number:
Molecular Weight:
89.09
Beilstein:
1720250
EC Number:
MDL number:
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

form

powder or crystals

color

white to off-white

mp

289 °C (dec.) (lit.)

SMILES string

CC(N)C(O)=O

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)

InChI key

QNAYBMKLOCPYGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

100, 500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service