Product Name
Yttrium(III) oxide 99.99% trace metals basis
Product Number
205168
Product Brand
ALDRICH
CAS Number
1314-36-9
Molecular Weight
225.81
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
TITRATION (KT) EDTA 0.1M77.6 - 79.9 % Y
PURITY≥ 99.99 % Based On Rare Earth Analysis
X-RAY DIFFRACTIONCONFORMS TO STRUCTURE
ICP ANALYSISConfirms Yttrium Component
METAL TRACE ANALYSIS (ICP)≤ 150 PPM (Trace Rare Earth Analysis)