Product Name
Nickel(II) chloride 98%
Product Number
339350
Product Brand
ALDRICH
CAS Number
7718-54-9
Molecular Weight
129.60
Test
Specification
APPEARANCE (COLOR)PALE TO DARK ORANGE
APPEARANCE (FORM)Powder
ARGENTOMETR. TITRATION≥ 53.3% CL
ICP ANALYSISCONFIRMS NICKEL COMPONENT