Product Name
Ammonia solution 4 M in methanol
Product Number
779423
Product Brand
ALDRICH
CAS Number
7664-41-7
Molecular Weight
17.03
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
TITRATION (METHOD)Back titration with NaOH
TITRATION (MOLARITY)3.6 - 4.4 mol/l
RECOMMENDED RETEST PERIOD36 MONTHS