Product Name
Sodium nitrate analytical standard
Product Number
15736
Product Brand
SIAL
CAS Number
7631-99-4
Molecular Weight
84.99
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
PURITY BASED ON TRACE METALS ANALYSIS≥ 99.995%
INFRARED SPECTRUMCONFORMS TO STRUCTURE
ICP ANALYSISConfirms Sodium Component
METAL TRACE ANALYSIS (ICP)Trace Metal Analysis ≤ 50.0 mg/kg
EXPIRY PERIOD36 MONTHS