Product Name
Chromoionophore I Selectophore
Product Number
27086
Product Brand
SIAL
CAS Number
125829-24-5
Molecular Weight
583.85
Test
Specification
APPEARANCE (COLOR)Conforms to Requirements
APPEARANCE (FORM)Powder or Crystals
PURITY (TLC AREA %)≥ 95.0 %
MELTING POINT98 - 106 C
CARBON CONTENT78.17 % (THEORY)
HYDROGEN CONTENT9.15 % (THEORY)
NITROGEN CONTENT7.20 % (THEORY)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD42 MONTHS