Product Name
Thiabendazole PESTANAL®, analytical standard
Product Number
45684
Product Brand
SIAL
CAS Number
148-79-8
Molecular Weight
201.25
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (GC AREA %)≥ 98.0 %
MELTING POINT298 - 304 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS