Product Name
2,6-Diisopropylphenol analytical standard
Product Number
56931
Product Brand
SIAL
CAS Number
2078-54-8
Molecular Weight
178.27
Test
Specification
APPEARANCE (COLOR)Colorless to Very Dark Yellow and Colorless to Very Dark Brown-Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
REFRACTIVE INDEX N20/D1.512 - 1.516
WATER≤ 0.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS