Product Name
N-Nitrosoanatabine analytical standard
Product Number
75281
Product Brand
SIAL
CAS Number
887407-16-1
Molecular Weight
189.21
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow and Colorless to Orange
APPEARANCE (FORM)Liquid or Solid
PURITY (HPLC AREA %)≥ 97.0 %
IDENTIFICATION (LC-MS)CORRESPONDS TO REQUIREMENTS
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS