Product Name
Hydrogen chloride solution 3 M in methanol, for GC derivatization, LiChropur
Product Number
90964
Product Brand
SIAL
CAS Number
7647-01-0
Molecular Weight
36.46
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
TITRATION (METHOD)ALKALIMETRIC
TITRATION (MOLARITY)2.7 - 3.3 mol/l
REMARKS ON GCSuitibility for GC derivatization complying
DENSITY D20/40.871 - 0.881
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD5 MONTHS