Product Name
Glyceryl tributyrate puriss., ≥98.5% (GC)
Product Number
91010
Product Brand
SIAL
CAS Number
60-01-5
Molecular Weight
302.36
Test
Specification
APPEARANCE (COLOR)Colorless to Almost Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.5 %
DENSITY D20/41.031 - 1.033
REFRACTIVE INDEX N20/D1.434 - 1.436
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1G IN 10ML CHLOROFORM
WATER≤ 0.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE