Product Name
Prilocaine hydrochloride meets USP testing specifications
Product Number
P3347
Product Brand
SIAL
CAS Number
1786-81-8
Molecular Weight
256.77
Test
Specification
IDENTITYPASS
MELTING RANGE166 DEG TO 169 DEG C
LOSS ON DRYINGNMT 0.3%
RESIDUE ON IGNITIONNMT 0.1%
HEAVY METALSNMT 0.002%
ASSAY99.0% TO 101.0% (DRY BASIS)
DOCUMENT # P3347/09/22/10/2