Product Name
Potassium hydroxide technical, ≥85%, powder
Product Number
06103
Product Brand
SIGALD
CAS Number
1310-58-3
Molecular Weight
56.11
Test
Specification
APPEARANCE (COLOR)White to Faint Beige
APPEARANCE (FORM)Powder
TITRATION (T) HCL 0.1M≥ 85 %
CONTENT OF CARBONATE (T)≤ 1.0 % K2CO3
METAL TRACE ANALYSIS (ICP)TOTAL IMPURITIES ≤ 0.002%-br-HEAVY METALS (AS PB)
IRON (ICP)≤ 20 mg/kg
TOTAL SULFUR AS SO4 (ICP)≤ 50 mg/kg
CHLORIDE (CL)≤ 1000 mg/kg