Product Name
Potassium hydroxide pellets, reag. Ph. Eur., ≥85%
Product Number
30603
Product Brand
SIGALD
CAS Number
1310-58-3
Molecular Weight
56.11
Test
Specification
TITRATION (T) HCL 0.1M≥ 85 %
CONTENT OF CARBONATE (T)K2CO3 ≤ 2 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
PROOF OF IDENTITYCONFORMS
ALUMINIUM≤ 0.001 %
CALCIUM≤ 0.001 %
IRON≤ 0.001 %
SODIUM≤ 1.0 %
NICKEL≤ 0.0005 %
CHLORIDE (CL)≤ 0.02 %
PHOSPHATE (PO4)≤ 0.01 %
SULFATE (SO4)≤ 0.02 %
RECOMMENDED RETEST PERIOD36 MONTHS