Product Name
Acetonitrile puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Product Number
33019
Product Brand
SIGALD
CAS Number
75-05-8
Molecular Weight
41.05
Test
Specification
APPEARANCE (COLOR)COMPLYING
APPEARANCE (FORM)COMPLYING
COLOR (IN APHA)≤ 10 APHA
PURITY (GC AREA %)≥ 99.5 %
DENSITY D20/40.781 - 0.783
REFRACTIVE INDEX N20/D1.3435 - 1.3445
BOILING POINT-
BOILING RANGE 80-82 DEG C: ≥ 95 %
WATER (COULOMETR.)≤ 0.1 %
RESIDUE (EVAPORATION)≤ 10 PPM
ACIDITYFREE ACID (AS CH3COOH): ≤ 0.001 %
ALKALINITYFREE ALKALI (AS NH3): ≤ 0.001 %
ALUMINIUM≤ 0.00005 %
BARIUM≤ 0.00001 %
BORON≤ 0.000002 %
CALCIUM≤ 0.00005 %
CADMIUM≤ 0.000005 %
COBALT≤ 0.000002 %
CHROMIUM≤ 0.000002 %
COPPER≤ 0.000002 %
IRON≤ 0.00001 %
MAGNESIUM≤ 0.00001 %
MANGANESE≤ 0.000002 %
NICKEL≤ 0.000002 %
LEAD≤ 0.00001 %
TIN≤ 0.00001 %
ZINC≤ 0.00001 %
RECOMMENDED RETEST PERIOD24 MONTHS