Product Name
MISSION® esiRNA targeting mouse Hsf1
Product Number
EMU040051
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
AppearanceColorless liquid
Concentration200 ng/µl
PCR Product - Gel ElectrophoresisPass
PCR Product - DNA SequencingPass
Concentration - OD260 or Fluorescence IntensityPass
Purified RNaseIII - Gel ElectrophoresisPass