Product Name
L-Glutamine solution 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Product Number
G7513
Product Brand
SIGMA
CAS Number
56-85-9
Molecular Weight
Test
Specification
Appearance (Turbidity)Clear
Appearance (Form)Solution
pH @ 25°C7.0 - 7.6
Osmolality- mOs/kg
Cell Culture Testing - MTTPass
Cell LineCell Line - Cell Types
Glutamine Concentration>= 200 mmol
SterilityPass
Endotoxin<= 0.5 EU/ml
Animal Derived Component FreeConforms