Product Name
Sodium chloride solution 0.9% in water, BioXtra, suitable for cell culture
Product Number
S8776
Product Brand
SIGMA
CAS Number
7647-14-5
Molecular Weight
58.44
Test
Specification
Appearance (Turbidity)Clear
Appearance (Colour)Colorless
Appearance (Form)Solution
pH-
Osmolality270 - 300 mOs/kg
Salt Toxicity TestPass
Cell LineCell Line - Cell Types
Key Element Conc - ICP (Sodium)3.3 - 3.7 g/l
SterilityPass
Endotoxin Level<= 1.0 EU/ml