Product Name
MONOCLONAL ANTI-BETA-ACTIN antibody produced in mouse clone 8H10D10, crude ascites, buffered aqueous solution
Product Number
SAB1305567
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
AppearanceReport Result
ConcentrationReport Result
Immunohistochemistry1:10~50
Western Blotting ( Cell Lysate)1:5000~20000
Flow CytometryNot determined
ELISANot determined
ImmunoprecipitationNot determined
Chromatin ImmunoprecipitationNot determined
ImmunofluorescenceNot determined