Product Name
p-Toluenesulfonic acid monohydrate ≥99% (calc. to H2O free subst.)
Product Number
27815
Product Brand
aldrich
CAS Number
6192-52-5
Molecular Weight
190.22
Test
Specification
ASSAY≥ 99 %
WATER8.0 - 12.0 %
RESIDUE ON IGNITION≤ 0.5 %
RECOMMENDED RETEST PERIOD42 MONTHS