Product Name
(S)-(6,6′-Dimethoxybiphenyl-2,2′-diyl)bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphine] ≥97%, optical purity ee: ≥99%
Product Number
29513
Product Brand
aldrich
CAS Number
910134-30-4
Molecular Weight
1151.56
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
PHOSPHORUS 31 NMR SPECTRUMCONFORMS TO STRUCTURE
REMARKSChemical Purity > 97%
EE > 99 %
RECOMMENDED RETEST PERIOD40 MONTHS