Product Name
Fraxin analytical standard
Product Number
80533
Product Brand
sial
CAS Number
524-30-1
Molecular Weight
370.31
Test
Specification
APPEARANCE (COLOR)White to Yellow
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95.0 %
WATER≤ 12.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS