Product Name
Triacontane analytical standard
Product Number
90270
Product Brand
sial
CAS Number
638-68-6
Molecular Weight
422.81
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals or Flakes
PURITY (GC AREA %)≥ 98.0 %
MELTING POINT64 - 67 C
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS