Product Name
Acetonitrile suitable for DNA synthesis, ≥99.9% (GC)
Product Number
34442
Product Brand
sigald
CAS Number
75-05-8
Molecular Weight
41.05
Test
Specification
TITRATABLE ACIDfree acid (as CH3COOH) ≤ 0.001 %
TITRATABLE BASEfree alkali (as NH3) ≤ 0.0001 %
PURITY (GC AREA %)≥ 99.9 %
WATER (COULOMETR.)≤ 0.001 %
RECOMMENDED RETEST PERIOD21 MONTHS