Product Name
Acetonitrile for preparative HPLC, ≥99.8% (GC)
Product Number
82063
Product Brand
sigald
CAS Number
75-05-8
Molecular Weight
41.05
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.8 %
WATER (COULOMETR.)≤ 0.1 %
RESIDUE (EVAPORATION)≤ 0.0001 %
ABSORBANCE (1)240NM ≤ 0.01
ABSORBANCE (2)220NM ≤ 0.3
EXPIRY PERIOD60 MONTHS