Product Name
DL-2-Aminoadipic acid ≥99%
Product Number
a0637
Product Brand
sigma
CAS Number
542-32-5
Molecular Weight
161.16
Test
Specification
APPEARANCEWHITE TO OFF-WHITE POWDER
SOLUBILITYCLEAR TO SLIGHTLY HAZY COLORLESS TO LIGHT YELLOW SOLUTION AT 50MG/ML IN 1M HYDROCHLORIC ACID
PURITY BY THIN LAYER CHROMATOGRAPHYMINIMUM 99%
RECOMMENDED RETEST6 YEARS
DOCUMENT # A0637/8/31/98/2