Merck
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit IgG fraction of antiserum, buffered aqueous solution

Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit

Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit IgG fraction of antiserum, buffered aqueous solution
Synonyms:
Anti-GFAP, Anti-FLJ45472, Anti-Intermediate filament protein

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Species Reactivity
Technique

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-2 of 2