RU
EN
Applications
Products
Services
Support
Home
Search Results
Bovine tendon powder
(2)
Bovine tendon powder
CAS Number:
9007-34-5
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
900723
Bovine tendon powder, neutral, type I collagen, suitable for biomedical research
Expand
View Pricing
900722
Bovine tendon powder, long fiber, Type I collagen, ≥60% collagen, suitable for biomedical research
Expand
View Pricing