Products
RU
EN
Products
Applications
Products
Services
Support
Home
Search Results
D-(+)-Biotin
(2)
D-(+)-Biotin
Synonyms:
D(+)-Biotin, D-(+)-Biotin
Empirical Formula (Hill Notation):
C
10
H
16
N
2
O
3
S
CAS Number:
58-85-5
Molecular Weight:
244.31
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
8.51209
D-(+)-Biotin, Novabiochem
®
Expand
View Pricing
2031
D-(+)-Biotin
Expand
View Pricing